Order Entry
Export
Orders LinkContactUsLinkComponent
1,3-Dihydroxynaphthalene 99+%

1,3-Dihydroxynaphthalene 99+%

1,3-Dihydroxynaphthalene 99+%
Certificates
1,3-Dihydroxynaphthalene 99+%

3 Options Available Below

Product Details & Documents
Synonyms:  Naphthalene-1,3-diol, Naphthalene-1,3-diol
  • CAS Number:132-86-5
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00003965
  • Molecular Weight:160.17 g/mol
  • Merck Index:12,06483
  • Boiling Pt:386 °C (1013 hPa)
  • Melting Pt:123…126 °C
  • Density:1,12 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:205-079-7

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Pink-orange to brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    123 to 126 °C
  • Titration with NaOH:
    ≥99.0 %
Product Family Options

Product Information