Order Entry
Export
ContactUsLinkComponent
2,7-Dimethoxynaphthalene
2,7-Dimethoxynaphthalene
Catalog # EHERC20775500
Supplier:  EHRENSTORFER
CAS Number:  
2,7-Dimethoxynaphthalene
Catalog # EHERC20775500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 mg
  • Seq:
    2275

Specifications

About this item

 :
 :188.23 g/mol
Boiling Pt:299 °C (1013 hPa)
Melting Pt:144 °C
EINECS:222-433-6
CAS Number:3469-26-9
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00004064