2,3-Diaminonaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:771-97-1
:
Formula:C₁₀H₁₀N₂
MDL Number:MFCD00004116
:158.2 g/mol
Boiling Pt:343 °C (1013 hPa)
Melting Pt:199 °C
Density:1.0968 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:212-241-0