Orders LinkContactUsLinkComponent
1,5-Diaminonaphthalene 97%
1,5-Diaminonaphthalene 97%
1,5-Diaminonaphthalene 97%
1,5-Diaminonaphthalene 97%
Purity: 97% CAS No.: 2243-62-1 Molecular Formula: C10H10N2 Formula Weight: 158.20 Melting Point: 189-192° Boiling Point: 200-210°/5mm UN No.: 3077 MDL No.: MFCD00004029
 :  1,5-naphthylenediamine, 1,5-Naphthalenediamine
  •  :2243-62-1
  •  :C₁₀H₁₀N₂
  •  :MFCD00004029
  •  :158.2 g/mol
  •  :200…210 °C (5 mmHg)
  •  :189…192 °C
  •  :1.4 g/cm³ (20 °C)
  •  :Ambient
  •  :218-817-8
  •  :3077
  •  :9,III
Product Family Options
  • Pack type
    Pk
  • Glass bottle for solids
    5 g
    1 * 5 g
    1 * 5 g - 28,20 €
    28,20 €
    1 * 5 g
       28,20 €
  • Glass bottle for solids
    25 g
    1 * 25 g
    1 * 25 g - 59,10 €
    59,10 €
    1 * 25 g
       59,10 €
  • Glass bottle for solids
    100 g
    1 * 100 g
    1 * 100 g - 191,00 €
    191,00 €
    1 * 100 g
       191,00 €