Order Entry
Export
Orders LinkContactUsLinkComponent
 

1,5-Diaminonaphthalene 97%

Health
Environment
1,5-Diaminonaphthalene 97%
Certificates
1,5-Diaminonaphthalene 97%

2 Options Available Below

Product Details & Documents
Synonyms:  1,5-naphthylenediamine, 1,5-Naphthalenediamine
  • CAS Number:2243-62-1
  • Formula:C₁₀H₁₀N₂
  • MDL Number:MFCD00004029
  • Molecular Weight:158.2 g/mol
  • Boiling Pt:381 °C (1013 hPa)
  • Melting Pt:187…190 °C
  • Density:1,4 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:218-817-8
  • UN:3077
  • ADR:9,III

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Grey to dark brown Powder
  • Infrared spectrum:
    Conforms
  • GC:
    ≥96.0 %
  • Titration with HClO4:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 49,00 €
    49,00 €
    1 * 25 g
       49,00 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 168,00 €
    168,00 €
    1 * 100 g
       168,00 €