1-Naphthyl isothiocyanate
Supplier: Thermo Scientific
Certificates
About this item
CAS Number:551-06-4
:
Formula:C₁₁H₇NS
MDL Number:MFCD00003882
:185.25 g/mol
Merck Index:12,06498
Boiling Pt:100 °C (0.2 mmHg)
Melting Pt:59 °C
Density:1.81 g/cm³ (20 °C)
EINECS:208-990-8
UN:2923
ADR:8,III