2,6-Naphthalenedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL523763-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:216.19 g/mol
Boiling Pt:422 °C (1013 hPa)
Melting Pt:366…368 °C
:M-Clarity™ quality level = MQ100
EINECS:214-527-0
CAS Number:1141-38-4
Formula:C₁₂H₈O₄
MDL Number:MFCD00004105