Order Entry
Export
ContactUsLinkComponent
1,4-Naphthalenedicarboxylic acid, Sigma-Aldrich®
Irritant
1,4-Naphthalenedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL333581-1G
Supplier:  Merck
CAS Number:  
Irritant
1,4-Naphthalenedicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL333581-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :216.19 g/mol
 :M-Clarity™ quality level = MQ100
EINECS:210-094-7
 :GHS07
CAS Number:605-70-9
Formula:C₁₂H₈O₄
MDL Number:MFCD00014312