Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dibromonaphthalene
1,4-Dibromonaphthalene ≥98%
1,4-Dibromonaphthalene ≥98%
:
Thermo Fisher Scientific
1,4-Dibromonaphthalene ≥98%
:
Thermo Fisher Scientific
:
83-53-4
:
:
C₁₀H₆Br₂
:
MFCD00041823
:
285.97 g/mol
:
310 °C (1013 hPa)
:
80…82 °C
:
Ambient
:
201-484-8
:
GHS07
msds_ALFAA18171
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years