Ammonium cerium(IV) sulphate dihydrate
Catalog # ROTH3338.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
Specifications
About this item
:
:632.55 g/mol
Melting Pt:130 °C
EINECS:231-567-4
CAS Number:10378-47-9
Formula:(NH₄)₂Ce(SO₄)₄·2H₂O
MDL Number:MFCD00149417