Order Entry
Export
Orders LinkContactUsLinkComponent
 

Lanthanum(III) nitrate

100 g
Catalog # ROTH8358.1
Oxidizing
Corrosive
Supplier:   Roth Carl
Lanthanum(III) nitrate
Catalog # ROTH8358.1
Supplier:  Roth Carl
CAS Number:  10099-59-9
Lanthanum(III) nitrate

100 g

CAS Number:  10099-59-9
Price
120,00 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:10099-59-9
  • Formula:LaN₃O₉
  • MDL Number:MFCD00011067
  • Molecular Weight:324.92
  • Storage Temperature:Refrigerator
  • EINECS:233-238-0
  • UN:1477
  • ADR:5.1,II
Specifications

Specifications

  • Pk:
    100 g

Specifications