Order Entry
Export
Orders LinkContactUsLinkComponent
2,5-Dimethylindole

2,5-Dimethylindole

Glass bottle for solids, 1 g
Catalog # MOLE22095440-1G
Supplier:   Molekula
2,5-Dimethylindole
Catalog # MOLE22095440-1G
Supplier:  Molekula
CAS Number:  1196-79-8
2,5-Dimethylindole

Glass bottle for solids, 1 g

CAS Number:  1196-79-8
Price
Inquire For Price
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  2,5-Dimethyl-1H-indole
  • CAS Number:1196-79-8
  • Formula:C₁₀H₁₁N
  • MDL Number:MFCD00005621
  • Molecular Weight:145.2 g/mol
  • Boiling Pt:282 °C (1013 hPa)
  • Melting Pt:116 °C
  • Density:1.08 g/cm³ (20 °C)
  • EINECS:214-816-1
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications