Order Entry
Export
ContactUsLinkComponent
2,3-Dimethylindole
2,3-Dimethylindole
Catalog # MOLEM69986194
Supplier:  Molekula
CAS Number:  
2,3-Dimethylindole
Catalog # MOLEM69986194
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :145.2 g/mol
Boiling Pt:285 °C (1013 hPa)
Melting Pt:111…112 °C
EINECS:202-076-2
CAS Number:91-55-4
Formula:C₁₀H₁₁N
MDL Number:MFCD00005617