1,2-Dimethylindole, Sigma-Aldrich®
Catalog # SIALD165603-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:145.2 g/mol
:M-Clarity™ quality level = MQ100
EINECS:212-877-9
CAS Number:875-79-6
Formula:C₁₀H₁₁N
MDL Number:MFCD00005802