Order Entry
Export
Orders LinkContactUsLinkComponent
 

7-Methylindole ≥98%

7-Methylindole ≥98%
Certificates
7-Methylindole ≥98%
7-Methylindole ≥98%

2 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 933-67-5 Molecular Formula: C9H9N Formula Weight: 131.18 Melting Point: 83-84° Boiling Point: 266° MDL No.: MFCD00005684

  • CAS Number:933-67-5
  • Formula:C₉H₉N
  • MDL Number:MFCD00005684
  • Molecular Weight:131.18 g/mol
  • Boiling Pt:266 °C
  • Melting Pt:83…84 °C
  • Density:1.0202 g/cm³ (99.6 °C)
  •  :Density: 1.0202 g/cm³ (99.6 °C)
  • Storage Temperature:Ambient
  • EINECS:213-270-1
Product Family Options

Product Information