5,6-Dimethoxyindole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14430-23-0
:
Formula:C₁₀H₁₁NO₂
MDL Number:MFCD00005675
:177.2 g/mol
Boiling Pt:198 °C (8 mmHg)
Melting Pt:154…160 °C
Storage Temperature:Ambient
EINECS:238-402-5