Order Entry
Export
Orders LinkContactUsLinkComponent
6-Fluoroindole

6-Fluoroindole

Glass bottle for solids, 1 g
Catalog # MOLE18394914-1G
Irritant
Supplier:   Molekula
6-Fluoroindole
Catalog # MOLE18394914-1G
Supplier:  Molekula
CAS Number:  399-51-9
6-Fluoroindole

Glass bottle for solids, 1 g

CAS Number:  399-51-9
Price
73,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  6-Fluoro-1H-indole
  • CAS Number:399-51-9
  • Formula:C₈H₆FN
  • MDL Number:MFCD00056933
  • Molecular Weight:135.14 g/mol
  • Melting Pt:72…77 °C
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications