Order Entry
Export
Orders LinkContactUsLinkComponent
6-Fluoroindole

6-Fluoroindole

Irritant
Supplier:   CHEMPUR
6-Fluoroindole
Supplier:  CHEMPUR
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  6-Fluoro-1H-indole
  • CAS Number:399-51-9
  • Formula:C₈H₆FN
  • MDL Number:MFCD00056933
  • Molecular Weight:135.14 g/mol
  • Melting Pt:72…77 °C
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information