Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ninhydrin

Irritant
Supplier:   Roth Carl
Ninhydrin
Supplier:  Roth Carl
Certificates
Ninhydrin

3 Options Available Below

Product Details & Documents
Synonyms:  Ninhydrin monohydrate, Trioxohydrindene monohydrate, 2,2-Dihydroxyindane-1,3-dione, 2,2-Dihydroxy-1,3-indanedione, Indanetrione hydrate, 2,2-Dihydroxyindan-1,3-dion, 1,2,3-Indantrione monohydrate
  • CAS Number:485-47-2
  • Formula:C₉H₆O₄
  • MDL Number:MFCD00003791
  • Molecular Weight:178.14 g/mol
  • Merck Index:13,06582
  • Boiling Pt:351 °C (1013 hPa)
  • Melting Pt:250…258 °C
  • Density:0.862 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-618-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 53,50 €
    53,50 €
    1 * 25 g
       53,50 €

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 134,00 €
    134,00 €
    1 * 100 g
       134,00 €

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 525,00 €
    525,00 €
    1 * 500 g
       525,00 €