Order Entry
Export
Orders LinkContactUsLinkComponent
1-Phenylimidazole

1-Phenylimidazole

Supplier:   CHEMPUR
1-Phenylimidazole
Supplier:  CHEMPUR
Certificates
1-Phenylimidazole

2 Options Available Below

Product Details & Documents
  • CAS Number:7164-98-9
  • Formula:C₉H₈N₂
  • MDL Number:MFCD00041204
  • Molecular Weight:144.18 g/mol
  • Boiling Pt:276 °C (1013 hPa)
  • Melting Pt:13 °C
  • Density:1.1397 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • UN:2384
  • ADR:3,II
  •  :Density: 1,1397 g/cm³ (20 °C)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g

This product has been blocked by your organization. Please contact your purchasing department for more information