1,1'-Thiocarbonyldiimidazole
Glass bottle for solids, 25 g
Catalog # MOLE15276557-25G
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 1,1'-Thiocarbonylbis(imidazole), 1,1'-Thiocarbonylbis-1H-imidazole
- CAS Number:6160-65-2
- Formula:C₇H₆N₄S
- MDL Number:MFCD00005289
- Molecular Weight:178.22 g/mol
- Melting Pt:105…106 °C
- Storage Temperature:Refrigerator
- EINECS:228-183-4
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



