1,1'-Carbonyldiimidazole
Glass bottle for solids, 100 g
Catalog # MOLEM10944806
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Di(1H-imidazol-1-yl)methanone, CDI Crosslinker, 1,1'-Carbonylbis(imidazole), 1,1'-Carbonylbis-1H-imidazole, N,N'-Carbonyldiimidazole, CDI
- CAS Number:530-62-1
- Formula:C₇H₆N₄O
- MDL Number:MFCD00005286
- Molecular Weight:162.15 g/mol
- Merck Index:14,01819
- Melting Pt:111…116 °C
- Storage Temperature:Ambient
- EINECS:208-488-9
- UN:3263
- ADR:8,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g



