Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4-Dimethylimidazole
2,4-Dimethylimidazole 90% tech.
2,4-Dimethylimidazole 90% tech.
:
Thermo Fisher Scientific
2,4-Dimethylimidazole 90% tech.
:
Thermo Fisher Scientific
:
930-62-1
:
:
C₅H₈N₂
:
MFCD00022365
:
96.13 g/mol
:
266 °C (733 mmHg)
:
85…87 °C
:
213-221-4
:
3263
:
8,III
:
GHS05
Appearance:
Yellow to orange Viscous liquid or mass
Infrared spectrum:
Conforms
GC:
≥88 %
ACRO21032