

1,2-Dimethylimidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1739-84-0
:
Formula:C₅H₈N₂
MDL Number:MFCD00005294
:96.13 g/mol
Boiling Pt:204 °C (1013 hPa)
Melting Pt:38 °C
Density:1.084 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:217-101-2
:GHS05
:GHS07