


Imidazole, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:288-32-4
:
Formula:C₃H₄N₂
MDL Number:MFCD00005183
Merck Index:13,04935
Boiling Pt:255 °C (1013 hPa)
Melting Pt:89…90 °C
Density:1.18 g/cm³ (20 °C)
Flash Pt:135 °C
Storage Temperature:Ambient
UN:2923
ADR:8,III
:68.08 g/mol
EINECS:206-019-2
:GHS05
:GHS08
:GHS07