4-Nitroimidazole
Supplier: CHEMPUR
Jump to:
About this item
Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
- CAS Number:3034-38-6
- Formula:C₃H₃N₃O₂
- MDL Number:MFCD00005196
- Molecular Weight:113.08 g/mol
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-224-7
2 Options Available Below
Product Details & Documents
Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole
- CAS Number:3034-38-6
- Formula:C₃H₃N₃O₂
- MDL Number:MFCD00005196
- Molecular Weight:113.08 g/mol
- Melting Pt:300 °C
- Storage Temperature:Ambient
- EINECS:221-224-7
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
This product has been blocked by your organization. Please contact your purchasing department for more information



