

Iodine monobromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:7789-33-5
:
Formula:IBr
MDL Number:MFCD00011353
:206.81 g/mol
Merck Index:14,5016
Boiling Pt:116 °C (dec.)
Melting Pt:50 °C (subl.)
Density:4.416
Storage Temperature:Ambient
EINECS:232-159-9
UN:3260
ADR:8,II
:GHS05
:GHS07