Order Entry
Export
ContactUsLinkComponent
 
 
4,4'-Dipyridyl disulphide 98%
 
undefined
 
4,4'-Dipyridyl disulphide 98%

 

  • Appearance:
    White to orange Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    75 to 79 °C
  • Titration with HClO4:
    ≥97.5 %