
2,2'-Dipyridyl disulphide 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2127-03-9
:
Formula:C₁₀H₈N₂S₂
MDL Number:MFCD00006287
:220.32 g/mol
Boiling Pt:348 °C (1013 hPa)
Melting Pt:56…60 °C
Flash Pt:210 °C
Storage Temperature:Refrigerator
EINECS:218-343-1
UN:3335
ADR:9,III
:GHS07
Specification Test Results
- Appearance:White to yellow to beige Powder
- Infrared spectrum:Conforms
- Melting point:55 to 60 °C
- GC:≥97.5 %