Sulphuryl chloride for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
Synonyms: Sulphuryl dichloride
CAS Number:7791-25-5
:
Formula:Cl₂O₂S
MDL Number:MFCD00011451
Boiling Pt:69.1
Melting Pt:–54 °C
Density:1.67 g/cm³ (20 °C)
Molecular Weight:134.97 g/mol
EINECS:232-245-6
UN:1834
ADR:6.1 (8) I
Merck Index:12,09153
:M-Clarity™ quality level = MQ200
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable: