
Ammonium thiocyanate
Supplier: Roth Carl
Certificates
About this item
CAS Number:1762-95-4
:
Formula:NH₄SCN
MDL Number:MFCD00011428
:76.12 g/mol
Merck Index:13,00564
Melting Pt:150 °C
Density:1.305 g/cm³ (20 °C)
EINECS:217-175-6
:GHS07
Storage Temperature:Ambient