Diammonium hydrogen orthophosphate
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
- CAS Number:7783-28-0
- Formula:(NH₄)₂HPO₄
- MDL Number:MFCD00010891
- Molecular Weight:132.06 g/mol
- Merck Index:13,00546
- Melting Pt:185 °C
- Density:1.619 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-987-8
- REACH:01-2119490974-22
2 Options Available Below
Product Details & Documents
Synonyms: Diammonium hydrogen orthophosphate, Ammonium hydrogen phosphate, di-Ammonium hydrogen phosphate, Ammonium Phosphate, Ammonium biphosphate, Fyrex, Ammonium phosphate dibasic, Ammonium monohydrogen phosphate, Diammonium hydrogen phosphate
- CAS Number:7783-28-0
- Formula:(NH₄)₂HPO₄
- MDL Number:MFCD00010891
- Molecular Weight:132.06 g/mol
- Merck Index:13,00546
- Melting Pt:185 °C
- Density:1.619 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:231-987-8
- REACH:01-2119490974-22



