Ammonium chloride
Supplier: Roth Carl
Jump to:
About this item
Synonyms: Ammonium muriate, Salmiac
- CAS Number:12125-02-9
- Formula:NH₄Cl
- MDL Number:MFCD00011420
- Molecular Weight:53.49 g/mol
- Merck Index:13,00510
- Melting Pt:335 °C
- Density:1.5256 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:235-186-4
3 Options Available Below
Product Details & Documents
Synonyms: Ammonium muriate, Salmiac
- CAS Number:12125-02-9
- Formula:NH₄Cl
- MDL Number:MFCD00011420
- Molecular Weight:53.49 g/mol
- Merck Index:13,00510
- Melting Pt:335 °C
- Density:1.5256 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:235-186-4



