Order Entry
Export
ContactUsLinkComponent
Tris(4-methylphenyl)phosphine, Sigma-Aldrich®
Tris(4-methylphenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL287830-5G
Supplier:  Merck
CAS Number:  
Tris(4-methylphenyl)phosphine, Sigma-Aldrich®
Catalog # SIAL287830-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :304.37 g/mol
Boiling Pt:400 °C (1013 hPa)
Melting Pt:144 °C
Density:1.11 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
EINECS:213-863-5
CAS Number:1038-95-5
Formula:C₂₁H₂₁P
MDL Number:MFCD00008542