

Dichloro(phenyl)phosphine ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:644-97-3
:
Formula:C₆H₅Cl₂P
MDL Number:MFCD00000528
:178.98 g/mol
Boiling Pt:99…101 °C (11 mmHg)
Melting Pt:–51 °C
Density:1.319
Flash Pt:102 °C (215 °F)
Storage Temperature:Ambient
EINECS:211-425-8
UN:2798
ADR:8,II
:GHS06
:GHS05