
Tricyclohexylphosphine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:2622-14-2
:
Formula:C₁₈H₃₃P
MDL Number:MFCD00003853
:280.43 g/mol
Boiling Pt:132…134 °C (0.02 mmHg)
Melting Pt:78 °C
Density:0.9 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:220-069-2
:GHS07
Storage Temperature:Freezer