Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tricyclohexylphosphine
Tricyclohexylphosphine, Technical Grade
Tricyclohexylphosphine, Technical Grade
:
Thermo Fisher Scientific
Tricyclohexylphosphine, Technical Grade
:
Thermo Fisher Scientific
:
2622-14-2
:
:
C₁₈H₃₃P
:
MFCD00003853
:
132…134 °C (0.02 mmHg)
:
76…78 °C
:
0.9 g/cm³ (20 °C)
:
280.43 g/mol
:
Ambient
:
220-069-2
:
GHS07
Environmentally Preferable:
msds_ALFA044854
Thermo Scientific Acroseal 25 Years