Choline chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:67-48-1
:
Formula:HOH₂CCH₂N(CH₃)₃Cl
MDL Number:MFCD00011721
:139.62 g/mol
Merck Index:13,02226
Melting Pt:244…247 °C
Density:∼1.205 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:200-655-4