Tetrapropylammonium bromide 98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 98+% CAS No.: 1941-30-6 Molecular Formula: (CH3(CH2)2)4NBr Formula Weight: 266.27 Melting Point: ca 270° dec. Beilstein Reference: 3567846
Synonyms: TPABr
- CAS Number:1941-30-6
- Formula:C₁₂H₂₈BrN
- MDL Number:MFCD00011840
- Molecular Weight:266.27 g/mol
- Melting Pt:∼ 270 °C (dec.)
- Storage Temperature:Ambient
- EINECS:217-727-6
3 Options Available Below
Product Details & Documents
Purity: 98+% CAS No.: 1941-30-6 Molecular Formula: (CH3(CH2)2)4NBr Formula Weight: 266.27 Melting Point: ca 270° dec. Beilstein Reference: 3567846
Synonyms: TPABr
- CAS Number:1941-30-6
- Formula:C₁₂H₂₈BrN
- MDL Number:MFCD00011840
- Molecular Weight:266.27 g/mol
- Melting Pt:∼ 270 °C (dec.)
- Storage Temperature:Ambient
- EINECS:217-727-6



