Order Entry
Export
ContactUsLinkComponent
Tetrabutylammonium bromide
Tetrabutylammonium bromide
Catalog # MOLE64093691-500G
Supplier:  Molekula
CAS Number:  
Tetrabutylammonium bromide
Catalog # MOLE64093691-500G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :322.37 g/mol
Melting Pt:100…103 °C
Density:1.148 g/cm³ (20 °C)
EINECS:216-699-2
CAS Number:1643-19-2
Formula:C₁₆H₃₆BrN
MDL Number:MFCD00011633