Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tetrabutylammonium bromide
Tetrabutylammonium bromide ≥98%
Tetrabutylammonium bromide ≥98%
:
Thermo Fisher Scientific
Tetrabutylammonium bromide ≥98%
:
Thermo Fisher Scientific
:
1643-19-2
:
:
C₁₆H₃₆BrN
:
MFCD00011633
:
322.37 g/mol
:
101…105 °C
:
1.15
:
216-699-2
:
GHS07
:
Ambient
msds_ALFAA10249
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years