Arsenic testing apparatus
Arsenic testing apparatus, complete
Catalog # HOFM40065.00
Supplier: HOFMANN GLASTECHNIK
Jump to:
Product Details & Documents
This apparatus is suitable for the specification of arsenic.
Certifications: According to German Pharmacopoeia.
Delivery information: The complete system consists of one Erlenmeyer flask 100 ml, socket NS 29/32, one top with cone NS 29/32 and spherical ground ball size 19/9, one top with spherical ground cup size 19/9 and one fork clamp for spherical ground joints size 19/9.
Specifications
- Description:Arsenic testing apparatus, complete