Order Entry
Export
Orders LinkContactUsLinkComponent
 

(Ethyl)triphenylphosphonium bromide

Glass bottle for solids, 100 g
Catalog # MOLEM66589242
Irritant
Supplier:   Molekula
(Ethyl)triphenylphosphonium bromide
Catalog # MOLEM66589242
Supplier:  Molekula
CAS Number:  1530-32-1
(Ethyl)triphenylphosphonium bromide

Glass bottle for solids, 100 g

CAS Number:  1530-32-1
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Ethyltriphenylphosphonium bromide
  • CAS Number:1530-32-1
  • Formula:C₂₀H₂₀BrP
  • MDL Number:MFCD00011838
  • Molecular Weight:371.26 g/mol
  • Melting Pt:205…207 °C
  • Storage Temperature:Ambient
  • EINECS:216-223-3
  • UN:2811
  • ADR:6.1,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications