Adapters for universal centrifuges
Supplier: Simport Scientific
Jump to:
About this item
These universal centrifuge adapters are made of acetal plastic.
- Reusable and freezable (–90 °C)
2 Options Available Below
Product Details & Documents
These universal centrifuge adapters are made of acetal plastic.
- Reusable and freezable (–90 °C)
Product Family Options
Product Information
- AvailabilityPrice
- SIMPT330AD15Adapters for 15 ml rotor bores or adapters
- SIMPT330AD50Adapters for 50 ml rotor bores or adapters