Order Entry
Export
Orders LinkContactUsLinkComponent
Triphenylsilane ≥99%

Triphenylsilane ≥99%

Triphenylsilane ≥99%
Certificates
Triphenylsilane ≥99%
Triphenylsilane ≥99%

2 Options Available Below

Product Details & Documents

Purity: 99% CAS No.: 789-25-3 Molecular Formula: (C6H5)3SiH Formula Weight: 260.41 Flash Point: 76°(169°F) Melting Point: 42-44° Boiling Point: 159-163°/3mm Beilstein Reference: 978182 MDL No.: MFCD00003003

  • CAS Number:789-25-3
  • Formula:C₁₈H₁₆Si
  • MDL Number:MFCD00003003
  • Molecular Weight:260.41 g/mol
  • Boiling Pt:159…163 °C (3 mmHg)
  • Melting Pt:42…44 °C
  • Density:1.152 g/cm³ (20 °C)
  • Flash Pt:76 °C (169 °F)
  • Storage Temperature:Ambient
  • EINECS:212-333-0
Product Family Options

Product Information