Diphenylsilane ≥97%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002
- CAS Number:775-12-2
- Formula:C₁₂H₁₂Si
- MDL Number:MFCD00003002
- Molecular Weight:184.31 g/mol
- Boiling Pt:95…97 °C (13 mmHg)
- Density:0.996
- Flash Pt:98 °C (208 °F)
- Storage Temperature:Ambient
- EINECS:212-271-4
3 Options Available Below
Product Details & Documents
Purity: 97% CAS No.: 775-12-2 Molecular Formula: C12H12Si Formula Weight: 184.32 Flash Point: 98°(208°F) Refractive Index: 1.5790 Boiling Point: 95-97°/13mm Beilstein Reference: 2935887 MDL No.: MFCD00003002
- CAS Number:775-12-2
- Formula:C₁₂H₁₂Si
- MDL Number:MFCD00003002
- Molecular Weight:184.31 g/mol
- Boiling Pt:95…97 °C (13 mmHg)
- Density:0.996
- Flash Pt:98 °C (208 °F)
- Storage Temperature:Ambient
- EINECS:212-271-4



