Order Entry
Export
Orders LinkContactUsLinkComponent
 

Diphenylsilane 97%

Irritant
Diphenylsilane 97%
Certificates
Diphenylsilane 97%

2 Options Available Below

Product Details & Documents
  • CAS Number:775-12-2
  • Formula:C₁₂H₁₂Si
  • MDL Number:MFCD00003002
  • Molecular Weight:184.31 g/mol
  • Boiling Pt:95…97 °C (13 mmHg)
  • Density:0.99 g/cm³
  • Flash Pt:98 °C
  • Storage Temperature:Ambient
  • EINECS:212-271-4

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5780 to 1.5800 (20°C, 589 nm)
  • GC:
    ≥96.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 50,20 €
    50,20 €
    1 * 5 g
       50,20 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 141,00 €
    141,00 €
    1 * 25 g
       141,00 €