Hexamethyldisilane
Glass bottle for solids, 25 g
Catalog # MOLE22895484-25G
Supplier: Molekula
Jump to:
Product Details & Documents
- CAS Number:1450-14-2
- Formula:C₆H₁₈Si₂
- MDL Number:MFCD00008258
- Molecular Weight:146.38 g/mol
- Boiling Pt:112 °C (1013 hPa)
- Melting Pt:13…15 °C
- Density:0.729 g/cm³ (20 °C)
- Storage Temperature:Refrigerator
- EINECS:215-911-0
- UN:1993
- ADR:3,II
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g



