Order Entry
Export
Orders LinkContactUsLinkComponent
Hexamethyldisilane 98+%

Hexamethyldisilane 98+%

Flammable
Hexamethyldisilane 98+%
Certificates
Hexamethyldisilane 98+%

3 Options Available Below

Product Details & Documents
  • CAS Number:1450-14-2
  • Formula:C₆H₁₈Si₂
  • MDL Number:MFCD00008258
  • Molecular Weight:146.38 g/mol
  • Boiling Pt:112 °C (1013 hPa)
  • Melting Pt:15 °C
  • Density:0.71 g/cm³
  • Flash Pt:11 °C
  • Storage Temperature:Ambient
  • EINECS:215-911-0
  • UN:1993
  • ADR:3,II

Documents

Specifications for Product Family

Specification Test Results

  • Appearance:
    Clear colorless Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.4211 to 1.4231 (20°C, 589 nm)
  • GC:
    ≥98.0 %
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle
    10 mL
    Log in for availability
    Log in for availability
    1 * 10 mL
    1 * 10 mL - 52,60 €
    52,60 €
    1 * 10 mL
       52,60 €

  • Glass bottle
    50 mL
    Log in for availability
    Log in for availability
    1 * 50 mL
    1 * 50 mL - 163,00 €
    163,00 €
    1 * 50 mL
       163,00 €

  • Glass bottle
    250 mL
    Log in for availability
    Log in for availability
    1 * 250 mL
    1 * 250 mL - 605,00 €
    605,00 €
    1 * 250 mL
       605,00 €