Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Diphenyl-1,1,2,2-tetramethyldisilane
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
:
Thermo Fisher Scientific
1,2-Diphenyl-1,1,2,2-tetramethyldisilane 95%
:
Thermo Fisher Scientific
:
1145-98-8
:
:
C₁₆H₂₂Si₂
:
MFCD00054786
:
270.52 g/mol
:
111 °C
:
34…37 °C
:
0.97 g/cm³ (20 °C)
:
Ambient
msds_ALFAH28705
Thermo Scientific Acroseal 25 Years